3-(Trifluoromethyl)benzyl mercaptan, tech. 90%
3-(Trifluoromethyl)benzyl mercaptan, tech. 90%
3-(Trifluoromethyl)benzyl mercaptan, tech. 90%
Thermo Scientific Chemicals

3-(Trifluoromethyl)benzyl mercaptan, tech. 90%

CAS: 25697-55-6 | C8H7F3S | 192.199 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L08155.03
also known as L08155-03
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.20
Save 7.55 (14%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 30-Jun-2026
54.20
Save 7.55 (14%)
Each
Chemical Identifiers
CAS25697-55-6
IUPAC Name[3-(trifluoromethyl)phenyl]methanethiol
Molecular FormulaC8H7F3S
InChI KeyCQIQWIMXCPTQPJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(CS)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)>88.0%
Appearance (Color)Clear, colorless
Refractive Index1.4860-1.4940 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only