Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate, 97%
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate, 97%
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate, 97%
Thermo Scientific Chemicals

Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate, 97%

CAS: 22232-54-8 | C7H10N2O2S | 186.229 g/mol
Have Questions?
Catalog NumberQuantity
L08218.14
also known as L08218-14
25 g
Catalog number L08218.14
also known as L08218-14
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS22232-54-8
IUPAC Nameethyl 3-methyl-2-sulfanylidene-2,3-dihydro-1H-imidazole-1-carboxylate
Molecular FormulaC7H10N2O2S
InChI KeyCFOYWRHIYXMDOT-UHFFFAOYSA-N
SMILESCCOC(=O)N1C=CN(C)C1=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
FormCrystals or powder or crystalline powder or fused solid
Melting Point (clear melt)119.0-128.0?C
Appearance (Color)White
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

Solubility
Soluble in water, ethanol, chloroform and acetone. Sightly soluble in water.

Notes
Light sensitive. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Chernov'yants, M. S.; Dolinkin, A. O.; Chernyshev, A. V.; Khohlov, E. V.; Golovanova, E. G. Interaction of antithyroid drugs with bovine serum albumin: Electrophoretic and fluorimetric study. J. Pharm. Sci. 2010, 99 (3), 1567-1573.
  2. Roy, G.; Das, D.; Mugesh, G. Bioinorganic chemistry aspects of the inhibition of thyroid hormone biosynthesis by anti-hyperthyroid drugs. Inorg. Chim. Acta 2007, 360 (1), 303-316.