N-Boc-D-phenylalanine, 98%
N-Boc-D-phenylalanine, 98%
N-Boc-D-phenylalanine, 98%
N-Boc-D-phenylalanine, 98%
Thermo Scientific Chemicals

N-Boc-D-phenylalanine, 98%

CAS: 18942-49-9 | C14H19NO4 | 265.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L08722.06
also known as L08722-06
Price (EUR)
64,20
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
64,20
Each
Add to cart
Chemical Identifiers
CAS18942-49-9
IUPAC Name(2R)-2-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoic acid
Molecular FormulaC14H19NO4
InChI KeyZYJPUMXJBDHSIF-LLVKDONJSA-N
SMILESCC(C)(C)OC(=O)N[C@H](CC1=CC=CC=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-24.5? ? 1.5? (c=1, ethanol)
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)82.5-88.5?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only