N,N-Dimethyldecanamide, 98%
N,N-Dimethyldecanamide, 98%
N,N-Dimethyldecanamide, 98%
N,N-Dimethyldecanamide, 98%
Thermo Scientific Chemicals

N,N-Dimethyldecanamide, 98%

CAS: 14433-76-2 | C12H25NO | 199.338 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 L08731.14
亦稱為 L08731-14
價格 (TWD)
-
申請報價
Quantity:
25 g
Request bulk or custom format
化學識別
CAS14433-76-2
IUPAC NameN,N-dimethyldecanamide
Molecular FormulaC12H25NO
InChI KeyHNXNKTMIVROLTK-UHFFFAOYSA-N
SMILESCCCCCCCCCC(=O)N(C)C
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥97.5%
Refractive Index1.4525-1.4565 @ 20?C
N,N-Dimethyldecanamide is used in pharmaceutical, agrochemicals, veterinary drug and intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethyldecanamide is used in pharmaceutical, agrochemicals, veterinary drug and intermediate for organic synthesis.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. W.J. Irwin; F.D. Sanderson; A. Li Wan Po. Percutaneous absorption of ibuprofen and naproxen: Effect of amide enhancers on transport through rat skin. International Journal of Pharmaceutics. 1990, 66 (1-3), 243-252.
  2. Yang-I Lin; S. A. LangJr.; Christina M. Seifert; R. G. Child; G. O. Morton; P. F. Fabio. Aldehyde Syntheses. Study of the preparation of 9,10-anthracenedicarboxaldehyde. J. Org. Chem. 1979, 44(25), 4701-4703.