Thermo Scientific Chemicals

6-Methylnicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: L08877.06
5 g, Each
Thermo Scientific Chemicals

6-Methylnicotinic acid, 99%, Thermo Scientific Chemicals

Catalog number: L08877.06
5 g, Each
Quantity
Catalog number: L08877.06
also known as L08877-06
Price (USD)
Price: 111.00
Online price: 94.65
Your price:
Quantity
-

Chemical Identifiers

CAS
3222-47-7
IUPAC Name
6-methylpyridine-3-carboxylate
Molecular Formula
C7H6NO2
InChI Key
RZOKQIPOABEQAM-UHFFFAOYSA-M
SMILES
CC1=CC=C(C=N1)C([O-])=O
Appearance (Color)
White to cream to pale brown
Form
Crystals or powder or crystalline powder or fused solid or flakes or granules
Assay (HPLC)
≥98.5%
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

6-Methylnicotinic acid is an intermediate of the drug etoricoxib (a non- steroidal anti-inflammatory drug for the treatment of arthritis and osteoarthritis). It is also used as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Methylnicotinic acid is an intermediate of the drug etoricoxib (a non- steroidal anti-inflammatory drug for the treatment of arthritis and osteoarthritis). It is also used as organic intermediates.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text