Thermo Scientific Chemicals

3-Methoxy-4-nitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L08891.14
25 g, Each
Thermo Scientific Chemicals

3-Methoxy-4-nitrobenzoic acid, 98+%, Thermo Scientific Chemicals

Catalog number: L08891.14
25 g, Each
Quantity
Catalog number: L08891.14
also known as L08891-14
Price (USD)
Price: 310.00
Online price: 263.65
Your price:
Quantity
-

Chemical Identifiers

CAS
5081-36-7
IUPAC Name
3-methoxy-4-nitrobenzoic acid
Molecular Formula
C8H7NO5
InChI Key
PWURRRRGLCVBMX-UHFFFAOYSA-N
SMILES
COC1=CC(=CC=C1[N+]([O-])=O)C(O)=O
Appearance (Color)
Cream to pale brown or pale yellow
Assay (HPLC)
≥98.0%
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥98.0 to ≤102.0%
Melting Point (clear melt)
230.0-236.0?C

Description

3-Methoxy-4-nitrobenzoic acid was used in detection of nitroaromatic compounds by CdSe- quantum dots capped with polyamidoamine dendrimer-1,12-diaminododecane core-generation 4 nanocomposites. It was used in the synthesis of vorozole derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methoxy-4-nitrobenzoic acid was used in detection of nitroaromatic compounds by CdSe- quantum dots capped with polyamidoamine dendrimer-1,12-diaminododecane core-generation 4 nanocomposites. It was used in the synthesis of vorozole derivatives.

Solubility
Insoluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text