4-n-Heptyloxybenzaldehyde, 97%
4-n-Heptyloxybenzaldehyde, 97%
4-n-Heptyloxybenzaldehyde, 97%
Thermo Scientific Chemicals

4-n-Heptyloxybenzaldehyde, 97%

CAS: 27893-41-0 | C14H20O2 | 220.312 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L08966.14
also known as L08966-14
Price (EUR)
191,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
191,00
Each
Chemical Identifiers
CAS27893-41-0
IUPAC Name4-(heptyloxy)benzaldehyde
Molecular FormulaC14H20O2
InChI KeyYBCKMIZXHKVONZ-UHFFFAOYSA-N
SMILESCCCCCCCOC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to white to yellow
Refractive Index1.5225-1.5275 @ 20?C
FormLiquid
Free acid (titration)≤1.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only