Isovaleramide, 98%
Isovaleramide, 98%
Isovaleramide, 98%
Thermo Scientific Chemicals

Isovaleramide, 98%

CAS: 541-46-8 | C5H11NO | 101.149 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L09072.06
also known as L09072-06
Price (USD)
60.65
Online Exclusive
66.80
Save 6.15 (9%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
60.65
Online Exclusive
66.80
Save 6.15 (9%)
Each
Chemical Identifiers
CAS541-46-8
IUPAC Name3-methylbutanamide
Molecular FormulaC5H11NO
InChI KeySANOUVWGPVYVAV-UHFFFAOYSA-N
SMILESCC(C)CC(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Colorless to white to cream to red
FormCrystals or powder or crystalline powder or flakes
Melting Point (clear melt)134.0-140.0?C
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only