1,4-Butane diisothiocyanate, 98%
1,4-Butane diisothiocyanate, 98%
1,4-Butane diisothiocyanate, 98%
Thermo Scientific Chemicals

1,4-Butane diisothiocyanate, 98%

CAS: 4430-51-7 | C6H8N2S2 | 172.264 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09234.06
also known as L09234-06
Price (EUR)
216,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
216,00
Each
Chemical Identifiers
CAS4430-51-7
IUPAC Name1,4-diisothiocyanatobutane
Molecular FormulaC6H8N2S2
InChI KeyRRSISCMPUAGVJN-UHFFFAOYSA-N
SMILESS=C=NCCCCN=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to pale brown
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)30.0-36.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only