Fluorescein isothiocyanate, isomer 1, 95%
Fluorescein isothiocyanate, isomer 1, 95%
Fluorescein isothiocyanate, isomer 1, 95%
Thermo Scientific Chemicals

Fluorescein isothiocyanate, isomer 1, 95%

CAS: 3326-32-7 | C21H11NO5S | 389.381 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
1 g
Catalog number L09319.03
also known as L09319-03
Price (USD)
332.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
332.00
Each
Chemical Identifiers
CAS3326-32-7
IUPAC Name3',6'-dihydroxy-5-isothiocyanato-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular FormulaC21H11NO5S
InChI KeyMHMNJMPURVTYEJ-UHFFFAOYSA-N
SMILESOC1=CC=C2C(OC3=CC(O)=CC=C3C22OC(=O)C3=CC(=CC=C23)N=C=S)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Appearance (Color)Orange to brown
Assay (HPLC)≥94.0%
Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescein isothiocyanate is used as fluorescent labeling reagent for proteins and for rapid identification of pathogens in fluorescent antibody technique. It is also used as a reagent for microsequencing of proteins and peptides (HPLC). On celite it is used for the labeling of fibrinogen.

Solubility
Slightly soluble in DMSO, ethanol, methanol DMF and acetone

Notes
Incompatible with strong oxidizing agents, strong bases and amines.
RUO – Research Use Only

General References:

  1. Reagent for fluorescent labelling of proteins: Anal. Biochem., 20, 178 (1968).
  2. Guilini, C.; Baehr, C.; Schaeffer, E.; Gizzi, P.; Rufi, F.; Haiech, J.; Weiss, E.; Bonnet, D.; Galzi, J. New Fluorescein Precursors for Live Bacteria Detection. Anal. Chem. 2015, 87 (17), 8858-8866.
  3. Chen, Y.; Yang, C.; Li, T.; Zhang, M.; Liu, Y.; Gauthier, M. A.; Zhao, Y.; Wu, C. The Interplay of Disulfide Bonds, α-Helicity, and Hydrophobic Interactions Leads to Ultrahigh Proteolytic Stability of Peptides. Biomacromolecules 2015, 16 (8), 2347-2355.