2-Bromo-5-fluorobenzyl bromide, 97%
2-Bromo-5-fluorobenzyl bromide, 97%
2-Bromo-5-fluorobenzyl bromide, 97%
2-Bromo-5-fluorobenzyl bromide, 97%
Thermo Scientific Chemicals

2-Bromo-5-fluorobenzyl bromide, 97%

CAS: 112399-50-5 | C7H5Br2F | 267.923 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09553.03
also known as L09553-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS112399-50-5
IUPAC Name1-bromo-2-(bromomethyl)-4-fluorobenzene
Molecular FormulaC7H5Br2F
InChI KeyCZLWYKAZAVYQIK-UHFFFAOYSA-N
SMILESFC1=CC(CBr)=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to cream to pale brown
FormCrystals or powder or crystalline powder or fused solid or clear liquid as melt
Melting Point (clear melt)30.0-40.0?C
Assay (GC)≥96.0%
Refractive Index1.5870-1.5920 @ 20?C
It is an important pharmaceutical intermediate. Alkylation of the β-amino ester 1a with 2-bromo-5-fluorobenzyl bromide yields Benzazepines as a white solid after silica gel chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important pharmaceutical intermediate. Alkylation of the β-amino ester 1a with 2-bromo-5-fluorobenzyl bromide yields Benzazepines as a white solid after silica gel chromatography.

Solubility
Difficult to mix in water.

Notes
Store in cool dry condition. Incompatible with strong oxidizing agents. Ensure ventilation. Protect from electrostatic charges.
RUO – Research Use Only
Valérie Declerck.; Patrice Ribière.; Yannig Nédellec.; Hassan Allouchi.; Jean Martinez.; Frédéric Lamaty. A Microwave-Assisted Heck Reaction in Poly(ethylene glycol) for the Synthesis of Benzazepines.European Journal of Organic Chemistry. 2007, 2007 (1),201-208 .