Thermo Scientific Chemicals

4-Aminobenzo-2,1,3-thiadiazole, 98%, Thermo Scientific Chemicals

Catalog number: L09678.06
5 g, Each
Thermo Scientific Chemicals

4-Aminobenzo-2,1,3-thiadiazole, 98%, Thermo Scientific Chemicals

Catalog number: L09678.06
5 g, Each
Quantity
Catalog number: L09678.06
also known as L09678-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
767-64-6
IUPAC Name
2,1,3-benzothiadiazol-4-amine
Molecular Formula
C6H5N3S
InChI Key
DRLGIZIAMHIQHL-UHFFFAOYSA-N
SMILES
NC1=CC=CC2=NSN=C12
Appearance (Color)
Yellow or Orange or Orange-yellow to brown
Form
Powder or crystalline powder or crystals
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Melting Point (clear melt)
66-70°C

Description

4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminobenzo-2,1,3-thiadiazole is used in a study to synthesize and evaluate the inhibitory activity of a series of substituted benzimidazoles and small benzothiadiazoles on rat liver methionine synthase. It was used as potential levelers for Cu plating in submicrometer trenches.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text