Fast Blue BB salt
Fast Blue BB salt
Fast Blue BB salt
Thermo Scientific Chemicals

Fast Blue BB salt

CAS: 5486-84-0 | C34H36Cl4N6O6Zn | 831.878 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
25 g
產品號碼 L09704.14
亦稱為 L09704-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS5486-84-0
IUPAC Name4-benzamido-2,5-diethoxybenzene-1-diazonium
Molecular FormulaC17H18N3O3
InChI KeyDKPBKNZQVUOHRQ-UHFFFAOYSA-O
SMILESCCOC1=CC([N+]#N)=C(OCC)C=C1NC(=O)C1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Yellow to green or dark brown
Formpowder
Assay (unspecified)> 80.0% dye content
Fast Blue BB salt is used as a general substrate for β-glycosidase. Two moles of 6-bromo-2-napthol-β-d-glucopyranoside are cleaved by β-glucosidases into 2 moles of d-glucose and 2 moles of 6-bromo-2-napthol. The 2 moles of 6-bromo-2-napthol formed are coupled with 1 mole of Fast Blue BB salt [4-benzoylamino-2,5-diethoxybenzenediazonium chloride hemi (zinc chloride) salt], resulting in 2 moles of the corresponding dye of 6-bromo-2-naphthol (reddish color). It is a commonly used stain to observe myelin under light microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fast Blue BB salt is used as a general substrate for β-glycosidase. Two moles of 6-bromo-2-napthol-β-d-glucopyranoside are cleaved by β-glucosidases into 2 moles of d-glucose and 2 moles of 6-bromo-2-napthol. The 2 moles of 6-bromo-2-napthol formed are coupled with 1 mole of Fast Blue BB salt [4-benzoylamino-2,5-diethoxybenzenediazonium chloride hemi (zinc chloride) salt], resulting in 2 moles of the corresponding dye of 6-bromo-2-naphthol (reddish color). It is a commonly used stain to observe myelin under light microscopy.

Solubility
Soluble in water.
RUO – Research Use Only
Helene Breitschopf.; Gerda Suchanek.; Robert M. Gould.; David R. Colman.; Hans Lassmann. In situ hybridization with digoxigenin-labeled probes: sensitive and reliable detection method applied to myelinating rat brain.Acta Neuropathologica. 1992, 84 (6),581-587 .