Thermo Scientific Chemicals

(S)-(+)-Prolinol, 98%, Thermo Scientific Chemicals

Catalog number: L09779.06
5 g, Each
Thermo Scientific Chemicals

(S)-(+)-Prolinol, 98%, Thermo Scientific Chemicals

Catalog number: L09779.06
5 g, Each
Quantity
Catalog number: L09779.06
also known as L09779-06
Price (JPY)

Chemical Identifiers

CAS
23356-96-9
IUPAC Name
(pyrrolidin-2-yl)methanol
Molecular Formula
C5H11NO
InChI Key
HVVNJUAVDAZWCB-UHFFFAOYNA-N
SMILES
OCC1CCCN1
Appearance (Color)
Clear colorless to yellow to green or blue
Form
Liquid or viscous liquid
Water Content (Karl Fischer Titration)
≤1%
Refractive Index
1.4830-1.4870 @ 20?C
Assay (GC)
≥97.5%

Description

It is an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical intermediate.

Solubility
Fully miscible in water. Soluble in chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text