4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%
4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%
4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%
Thermo Scientific Chemicals

4-Chloro-2-(trifluoromethyl)phenyl isocyanate, 97%

CAS: 16588-69-5 | C8H3ClF3NO | 221.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L09850.06
also known as L09850-06
Price (USD)
109.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
109.00
Each
Chemical Identifiers
CAS16588-69-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive Index1.5000-1.5050 @ 20?C
FormLiquid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only