Thermo Scientific Chemicals

4,4-Dimethyl-2-pentyne, 97+%, Thermo Scientific Chemicals

Catalog number: L10210.06
5 g, Each
Thermo Scientific Chemicals

4,4-Dimethyl-2-pentyne, 97+%, Thermo Scientific Chemicals

Catalog number: L10210.06
5 g, Each
Quantity
Catalog number: L10210.06
also known as L10210-06
Price (USD)
Price: 188.00
Online price: 159.65
Your price:
Quantity
-

Chemical Identifiers

CAS
999-78-0
Assay from Suppliers CofA
≥97.0% (GC, U.S. specification)
Comment
Specification differs for U.S. and non-U.S. material where indicated
Refractive Index
1.4050-1.4100 @ 20°C (non-U.S. specification)
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid

Description

4,4-Dimethyl-2-pentyne is used in organometallic reactions for regioselectivity of its insertion into the Pd-C bond of cyclopalladated complexes. It is also used to prepare pyrimidine-dione by reacting with isocyanate in the presence of nickel/ N-heterocyclic carbene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Dimethyl-2-pentyne is used in organometallic reactions for regioselectivity of its insertion into the Pd-C bond of cyclopalladated complexes. It is also used to prepare pyrimidine-dione by reacting with isocyanate in the presence of nickel/ N-heterocyclic carbene.

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text