Tantalum(V) ethoxide, 99+%
Tantalum(V) ethoxide, 99+%
Tantalum(V) ethoxide, 99+%
Thermo Scientific Chemicals

Tantalum(V) ethoxide, 99+%

CAS: 6074-84-6 | C10H25O5Ta | 406.25 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
產品號碼 L10288.14
亦稱為 L10288-14
價格 (TWD)
-
Quantity:
25 g
Request bulk or custom format
化學識別
CAS6074-84-6
IUPAC Namepentaethoxytantalum
Molecular FormulaC10H25O5Ta
InChI KeyHSXKFDGTKKAEHL-UHFFFAOYSA-N
SMILESCCO[Ta](OCC)(OCC)(OCC)OCC
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to pale yellow
FormLiquid (as melt)
Assay (special)≥98.5% (based on tantalum EA)
Elemental AnalysisTantalum, Ta: 44.10 to 44.99%
Elemental Analysis (typical)Niobium, Nb <80 ppm
檢視更多
Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tantalum(V) ethoxide acts as a precursor used in the preparation of ultra thin films of tantalum oxide and other tantalum containing films, which finds application in semiconductors. It is also used to prepare tantalum oxide nanoparticles for the imaging cartilage.

Solubility
Soluble in organic solvents.

Notes
Moisture sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Hildebrandt, N. C.; Soldat, J.; Marschall, R. Layered Perovskite Nanofibers via Electrospinning for Overall Water Splitting. Small 2015, 11 (17), 2051-2057.
  2. Morris, R. S.; Lavoie, G. G. Strong π-interactions between tantalum and apical ligands in square pyramidal complexes supported by a rigid tetradentate spectator ligand. Inorg. Chim. Acta 2015, 435, 1-6.
  3. Sfirloaga, P.; Miron, I.; Malaescu, I.; Marin, C. N.; Ianasi, C.; Vlazan, P. Structural and physical properties of undoped and Ag-doped NaTaO3 synthesized at low temperature. Mater. Sci. Semicond. Process. 2015, 39, 721-725.