Thermo Scientific Chemicals

Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals

Catalog number: L10386.22
100 g, Each
Thermo Scientific Chemicals

Diisopropyl azodicarboxylate, 94%, Thermo Scientific Chemicals

Catalog number: L10386.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L10386.22
also known as L10386-22
Price (USD)
Price: 112.00
Your price:
Quantity
-

Chemical Identifiers

CAS
2446-83-5
IUPAC Name
N-{[(propan-2-yloxy)carbonyl]imino}(propan-2-yloxy)formamide
Molecular Formula
C8H14N2O4
InChI Key
VVWRJUBEIPHGQF-UHFFFAOYSA-N
SMILES
CC(C)OC(=O)N=NC(=O)OC(C)C
Appearance (Color)
Clear, orange to reddish brown
Form
Liquid
Assay (GC)
≥ 92.0%

Description

Diisopropyl azodicarboxylate is used as an important reagent in the production of many organic compounds. It is used in association with triphenyl phosphine in Mitsunobu reaction of alcohols, acids and amides. It acts as reactant in the preparation of chromenes resembling classical cannabinoids, norbornene-based guanidine-rich polymers and acceptor-donor-acceptor organic dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diisopropyl azodicarboxylate is used as an important reagent in the production of many organic compounds. It is used in association with triphenyl phosphine in Mitsunobu reaction of alcohols, acids and amides. It acts as reactant in the preparation of chromenes resembling classical cannabinoids, norbornene-based guanidine-rich polymers and acceptor-donor-acceptor organic dyes.

Solubility
Immiscible with water.

Notes
Store in cool place. Light sensitive. Incompatible with strong oxidizing agents, strong bases, alcohols and metallic salts. Decomposes violently at or above: 100 0C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text