trans-3-Hexen-1-ol, 97%
trans-3-Hexen-1-ol, 97%
trans-3-Hexen-1-ol, 97%
Thermo Scientific Chemicals

trans-3-Hexen-1-ol, 97%

CAS: 928-97-2 | C6H12O | 100.161 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10541.06
also known as L10541-06
Price (EUR)
62,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
62,10
Each
Chemical Identifiers
CAS928-97-2
IUPAC Name(3E)-hex-3-en-1-ol
Molecular FormulaC6H12O
InChI KeyUFLHIIWVXFIJGU-ONEGZZNKSA-N
SMILESCCC=CCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Assay (GC)≥96.0%
Appearance (Color)Clear colorless to yellow
Refractive Index1.4370-1.4435 @ 20?C
trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-3-Hexen-1-ol stimulates the antennae of male H. cunea moths. It is also used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Rui Tang; Jin Ping Zhang; Zhong Ning Zhang. Electrophysiological and behavioral responses of male fall webworm moths (Hyphantria cunea) to Herbivory-induced mulberry (Morus alba) leaf volatiles. PLoS ONE. 2012, 7, (11),e49256
  2. Shigehiro Namiki; Satoshi Iwabuchi; Ryohei Kanzaki. Representation of a mixture of pheromone and host plant odor by antennal lobe projection neurons of the silkmoth Bombyx mori. Journal of Comparative Physiology, A. 2008, 194, (5),501-515