Vitamin K{1}
Vitamin K{1}
Vitamin K{1}
Thermo Scientific Chemicals

Vitamin K{1}

Important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver. | CAS: 84-80-0 | C31H46O2 | 450.707 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L10575.06
also known as L10575-06
Price (EUR)
174,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
174,00
Each
Chemical Identifiers
CAS84-80-0
IUPAC Name2-methyl-3-[(2E)-3,7,11,15-tetramethylhexadec-2-en-1-yl]-1,4-dihydronaphthalene-1,4-dione
Molecular FormulaC31H46O2
InChI KeyMBWXNTAXLNYFJB-LKUDQCMENA-N
SMILESCC(C)CCCC(C)CCCC(C)CCCC(C)=CCC1=C(C)C(=O)C2=CC=CC=C2C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow or orange to red
FormViscous liquid
Assay from Suppliers CofA≥97.5% (cis + trans)
Comment5-25% cis isomer
Identification (FTIR)Conforms
View more
Vitamin K{1} is an important dietary source found in green plants, algae and photosynthetic bacteria. It is used as a vitamin (prothrombogenic) and veterinary antidote for dicoumarol poisoning. It is an essential vitamin for the synthesis of coagulation factors II, VII, IX, and X in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only