Thiophene-2-carboxaldoxime, 98+%
Thiophene-2-carboxaldoxime, 98+%
Thiophene-2-carboxaldoxime, 98+%
Thermo Scientific Chemicals

Thiophene-2-carboxaldoxime, 98+%

CAS: 29683-84-9 | C5H5NOS | 127.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L10961.14
also known as L10961-14
Price (TWD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS29683-84-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)white to pale cream
Assay (HPLC)≥98.0%
FormCrystals or crystalline powder
Melting Point132-138?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only