Thermo Scientific Chemicals

Norbornene, 99%, Thermo Scientific Chemicals

Catalog number: L11103.36
500 g, Each
Thermo Scientific Chemicals

Norbornene, 99%, Thermo Scientific Chemicals

Catalog number: L11103.36
500 g, Each
Quantity
Catalog number: L11103.36
also known as L11103-36
Price (USD)
Price: 121.00
Online price: 102.65
Your price:
Quantity
-

Chemical Identifiers

CAS
498-66-8
IUPAC Name
bicyclo[2.2.1]hept-2-ene
Molecular Formula
C7H10
InChI Key
JFNLZVQOOSMTJK-UHFFFAOYNA-N
SMILES
C1CC2CC1C=C2
Comment
May contain up to 0.5% toluene
Appearance (Color)
White or translucent
Form
Crystals or powder or crystalline powder or fused solid, clear as melt
Assay from Supplier's CofA
≥98.5%

Description

Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs' catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Norbornene is used as a monomer and as an intermediate in organic synthesis. It is also used to prepare pharmaceutical intermediates and specialty fragrances. It reacts with ethylene to get cyclic olefin copolymer. Further, it is used in transition metal catalysis to affect migration of electrophilic transition metals. It acts as a monomer in ring-opening metathesis polymerizations with the Grubbs′ catalyst. Its polymer, polynorbornene is used in the rubber industry for anti-vibration, anti-impact in personal protective equipment and grip improvement in toy tires, racing tires and transmission system.

Solubility
Soluble in ethanol, ether and most organic solvents. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text