1-Heptene, 98+%
1-Heptene, 98+%
1-Heptene, 98+%
Thermo Scientific Chemicals

1-Heptene, 98+%

CAS: 592-76-7 | C7H14 | 98.189 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Catalog number L11487.AE
also known as L11487-AE
Price (EUR)
186,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
186,00
Each
Chemical Identifiers
CAS592-76-7
IUPAC Namehept-1-ene
Molecular FormulaC7H14
InChI KeyZGEGCLOFRBLKSE-UHFFFAOYSA-N
SMILESCCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
CommentMaterial sourced in UK and US
FormLiquid
Assay (GC)≥98.0%
Refractive Index1.3975-1.4015 @ 20?C (UK Sourced Material)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only