2,4,5-Trichlorophenyl isothiocyanate, 98%
2,4,5-Trichlorophenyl isothiocyanate, 98%
2,4,5-Trichlorophenyl isothiocyanate, 98%
Thermo Scientific Chemicals

2,4,5-Trichlorophenyl isothiocyanate, 98%

CAS: 23165-46-0 | C7H2Cl3NS | 238.51 g/mol
製品番号(カタログ番号) L11551.03
または、製品番号L11551-03
価格(JPY)
-
数量:
1 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS23165-46-0
IUPAC Name1,2,4-trichloro-5-isothiocyanatobenzene
Molecular FormulaC7H2Cl3NS
InChI KeyPJLRSYLEFZNICX-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=C(C=C1Cl)N=C=S
さらに表示
Appearance (Color)White to cream
FormPowder
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only