Thermo Scientific Chemicals

4-Diethylaminosalicylaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: L11659.14
25 g, Each
Thermo Scientific Chemicals

4-Diethylaminosalicylaldehyde, 99%, Thermo Scientific Chemicals

Catalog number: L11659.14
25 g, Each
Quantity
Catalog number: L11659.14
also known as L11659-14
Price (USD)
Price: 70.20
Online price: 59.65
Your price:
Quantity
-

Chemical Identifiers

CAS
17754-90-4
IUPAC Name
4-(diethylamino)-2-hydroxybenzaldehyde
Molecular Formula
C11H15NO2
InChI Key
XFVZSRRZZNLWBW-UHFFFAOYSA-N
SMILES
CCN(CC)C1=CC=C(C=O)C(O)=C1
Form
Powder
Melting Point (clear melt)
59-64°C
Appearance (Color)
Cream to yellow or pink to brown to dark purple or pink/purple
Assay (GC)
≥96.0%

Description

4-Diethylaminosalicylaldehyde is used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Diethylaminosalicylaldehyde is used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.

Solubility
Insoluble in water.

Notes
It is sensitive to air. Store at room temperature. Incompatible with strong bases and strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text