4-Isopropylbenzyl chloride, 95%
4-Isopropylbenzyl chloride, 95%
4-Isopropylbenzyl chloride, 95%
Thermo Scientific Chemicals

4-Isopropylbenzyl chloride, 95%

CAS: 2051-18-5 | C10H13Cl | 168.664 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11839.06
also known as L11839-06
Price (EUR)
54,50
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
54,50
Each
Chemical Identifiers
CAS2051-18-5
IUPAC Name1-(chloromethyl)-4-(propan-2-yl)benzene
Molecular FormulaC10H13Cl
InChI KeyCYAKWEQUWJAHLW-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(CCl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥94.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only