4-(Trifluoromethyl)phenyl isothiocyanate, 98+%
4-(Trifluoromethyl)phenyl isothiocyanate, 98+%
4-(Trifluoromethyl)phenyl isothiocyanate, 98+%
Thermo Scientific Chemicals

4-(Trifluoromethyl)phenyl isothiocyanate, 98+%

CAS: 1645-65-4 | C8H4F3NS | 203.182 g/mol
Have Questions?
Catalog NumberQuantity
L11890.06
also known as L11890-06
5 g
Catalog number L11890.06
also known as L11890-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1645-65-4
IUPAC Name1-isothiocyanato-4-(trifluoromethyl)benzene
Molecular FormulaC8H4F3NS
InChI KeyDQEVDFQAYLIBRD-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)N=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or Fused solid
Identification (FTIR)Conforms
Appearance (Color)White
Melting Point (clear melt)38-44?C
Assay (GC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only