Thiazolyl Blue tetrazolium bromide, 98%
Thiazolyl Blue tetrazolium bromide, 98%
Thiazolyl Blue tetrazolium bromide, 98%
Thermo Scientific Chemicals

Thiazolyl Blue tetrazolium bromide, 98%

CAS: 298-93-1 | C18H16BrN5S | 414.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L11939.03
also known as L11939-03
Price (EUR)
46,65
Online exclusive
51,40
Save 4,75 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
46,65
Online exclusive
51,40
Save 4,75 (9%)
Each
Chemical Identifiers
CAS298-93-1
IUPAC Name2-(4,5-dimethyl-1,3-thiazol-2-yl)-3,5-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium bromide
Molecular FormulaC18H16BrN5S
InChI KeyAZKSAVLVSZKNRD-UHFFFAOYSA-M
SMILES[Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Identification (FTIR)Conforms
Assay (Argentometric Titration)≥97.5 to ≤102.5%
Appearance (Color)White to yellow
Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells.Thiazolyl Blue tetrazolium bromide is a dye used in measurement of cell proliferation. It is used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. As an electron acceptor, it is utilized for studying nicotinamide adenine dinucleotide phosphate (NADP)-dependent dehydrogenases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in measurement of cell proliferation. Produces a yellowish solution that is converted to dark blue, water-insoluble MTT formazan by mitochondrial dehydrogenases of living cells.Thiazolyl Blue tetrazolium bromide is a dye used in measurement of cell proliferation. It is used as a direct indicator of cytotoxicity (such as for screening cancer drugs) and apoptosis. As an electron acceptor, it is utilized for studying nicotinamide adenine dinucleotide phosphate (NADP)-dependent dehydrogenases.

Solubility
Soluble in water, ethanol, 2-methoxyethanol and phosphate buffered saline.

Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ma, W.; Xu, A.; Ying, J.; Li, B.; Jin, Y. Biodegradable Core-Shell Copolymer-Phospholipid Nanoparticles for Combination Chemotherapy: An In Vitro Study. J. Biomed. Nanotechnol. 2015, 11 (7), 1193-1200.
  2. Wei, D.; Chen, T.; Yan, M.; Zhao, W.; Li, F.; Cheng, W.; Yuan, L. Synthesis, characterization, antioxidant activity and neuroprotective effects of selenium polysaccharide from Radix hedysari. Carbohydr. Polym. 2015, 125, 161-168.