Thermo Scientific Chemicals

Cyclopropylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L11950.14
25 g, Each
Thermo Scientific Chemicals

Cyclopropylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog number: L11950.14
25 g, Each
Quantity
Catalog number: L11950.14
also known as L11950-14
Price (USD)
Price: 271.00
Online price: 230.65
Your price:
Quantity
-

Chemical Identifiers

CAS
6542-60-5
IUPAC Name
2-cyclopropylacetonitrile
Molecular Formula
C5H7N
InChI Key
FAUQRRGKJKMEIW-UHFFFAOYSA-N
SMILES
N#CCC1CC1
Form
Liquid
Refractive Index
1.4220-1.4270 @ 20?C
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%

Description

Cyclopropylacetonitrile is used in the preparation of cyclopropyl-2 acetophenone and cyclopropylacetyl chloride. It acts as an organic intermediate in organic synthesis. It is also employed in the preparation of agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopropylacetonitrile is used in the preparation of cyclopropyl-2 acetophenone and cyclopropylacetyl chloride. It acts as an organic intermediate in organic synthesis. It is also employed in the preparation of agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Miscible with water, methylene chloride and alcohols.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text