1-(4-Metoxifenil)etanol, 95 %, Thermo Scientific Chemicals
1-(4-Metoxifenil)etanol, 95 %, Thermo Scientific Chemicals
1-(4-Metoxifenil)etanol, 95 %, Thermo Scientific Chemicals
1-(4-Metoxifenil)etanol, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(4-Metoxifenil)etanol, 95 %, Thermo Scientific Chemicals

CAS: 3319-15-1 | C9H12O2 | 152.19 g/mol
Have Questions?
Número de catálogoCantidad
L12062.14
también denominado L12062-14
25 g
Número de catálogo L12062.14
también denominado L12062-14
Precio (USD)
-
Cantidad:
25 g
Pedido a granel o personalizado
Identificadores químicos
CAS3319-15-1
IUPAC Name1-(4-methoxyphenyl)ethan-1-ol
Molecular FormulaC9H12O2
InChI KeyIUUULXXWNYKJSL-UHFFFAOYNA-N
SMILESCOC1=CC=C(C=C1)C(C)O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Assay (GC)≥94.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.5300-1.5360 @ 20?C
FormLiquid
Identification (FTIR)Conforms
1-(4-Methoxyphenyl)ethanol is used to study the steady-state and nanosecond, laser-flash photolysis. It is used to produce 4-(1-chloro-ethyl)-anisole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El 1-(4-metoxifenil)etanol se utiliza para estudiar la fotolisis de estado estable, nanosegundo y láser-flash. Se utiliza para producir 4-(1-cloro-etil)-anisol.

Solubilidad
No miscible en agua.

Notas
Almacenar en un lugar fresco y seco y en un recipiente bien sellado. Incompatible con los agentes oxidantes.
RUO – Research Use Only

General References:

  1. Tiziana Del Giacco; Annalisa Faltoni; Fausto Elisei. Anomalous reactivity of radical cations produced by photosensitized oxidation of 4-methoxybenzyl alcohol derivatives: role of the sensitizer. Physical Chemistry Chemical Physics.2008, 10, (1), 200-210.
  2. Xiao-Hong Chen; Ping Wei; Xiao-Ting Wang; Min-Hua Zong; Wen-Yong Lou. A novel carbonyl reductase with anti-Prelog stereospecificity from Acetobacter sp. CCTCC M209061: purification and characterization. PLoS ONE.2014, 9, (4), e94543.