1,2:5,6-Di-O-isopropylidene-D-mannitol
1,2:5,6-Di-O-isopropylidene-D-mannitol
1,2:5,6-Di-O-isopropylidene-D-mannitol
Thermo Scientific Chemicals

1,2:5,6-Di-O-isopropylidene-D-mannitol

CAS: 1707-77-3 | C12H22O6 | 262.302 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L12127.14
also known as L12127-14
Price (EUR)
171,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
171,00
Each
Chemical Identifiers
CAS1707-77-3
IUPAC Name1,2-bis(2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
Molecular FormulaC12H22O6
InChI KeyODYBCPSCYHAGHA-UHFFFAOYNA-N
SMILESCC1(C)OCC(O1)C(O)C(O)C1COC(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder or crystalline powder
Assay (GC)≥96%
Water Content (Karl Fischer Titration)≤1%
Melting point118-124°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only