Diclorodimetilsilano, 99 %, Thermo Scientific Chemicals
Diclorodimetilsilano, 99 %, Thermo Scientific Chemicals
Diclorodimetilsilano, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Diclorodimetilsilano, 99 %, Thermo Scientific Chemicals

CAS: 75-78-5 | C2H6Cl2Si | 129.055 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
100 g
500 g
1000 g
Número de catálogo L12133.36
también denominado L12133-36
Precio (EUR)
66,20
Each
Cantidad:
500 g
Pedido a granel o personalizado
Precio (EUR)
66,20
Each
Identificadores químicos
CAS75-78-5
IUPAC Namedichlorodimethylsilane
Molecular FormulaC2H6Cl2Si
InChI KeyLIKFHECYJZWXFJ-UHFFFAOYSA-N
SMILESC[Si](C)(Cl)Cl
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormLiquid
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4025-1.4065 @ 20?C
Assay (GC)≥98.0%
Dichlorodimethylsilane is used to prepare a resin bound siloxane with tertiary alcohols and it is also used as a reagent for synthesis of optically active ansa-mettallocene polymerization catalysts. It acts as a precursor to silicone and polysilane compounds. It is also used in the glass coating to protect it from micro particles. It is involved in the preparation of resin bound siloxane with reactivity towards tertiary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
El diclorodimetilsilano se utiliza para preparar un siloxano enlazado a resina con alcoholes terciarios y también se utiliza como reactivo para la síntesis de catalizadores de polimerización de ansa-metaloceno ópticamente activos. Actúa como precursor de los compuestos de silicona y polisilano. También se utiliza en el revestimiento de vidrio para protegerlo de las micropartículas. Participa en la preparación del siloxano enlazado a resina con reactividad hacia los alcoholes terciarios.

Solubilidad
Inmiscible con agua.

Notas
Sensible a la humedad. Almacenar en lugar fresco. Incompatible con alcoholes, aminas y bases fuertes.
RUO – Research Use Only

General References:

  1. Silicone intermediate and silylating agent, forming dimethylsilylene derivatives of diols. The main limitation of these is their extreme susceptibilty to hydrolysis. See Appendix 4.
  2. For use in combination with triflic acid to generate dimethyldisilyl triflate, and its use in synthesis, see N,N'-Diacetyl hydrazine, A13393 .
  3. Kim, Y. R.; Lai, S. C. S.; McKelvey, K.; Zhang, G.; Perry, D.; Miller, T. S.; Unwin, P. R. Nucleation and aggregative growth of palladium nanoparticles on carbon electrodes: experiment and kinetic model. J. Phys. Chem. C 2015, 119 (30), 17389-17397.
  4. Wang, Z.; Gong, X.; Ngai, T. Measurements of Long-Range Interactions between Protein-Functionalized Surfaces by Total Internal Reflection Microscopy. Langmuir. 2015, 31 (10), 3101-3107.