(1-Decyl)trimethylammonium bromide, 98%
(1-Decyl)trimethylammonium bromide, 98%
(1-Decyl)trimethylammonium bromide, 98%
Thermo Scientific Chemicals

(1-Decyl)trimethylammonium bromide, 98%

CAS: 2082-84-0 | C13H30BrN | 280.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L12144.18
also known as L12144-18
Price (EUR)
149,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
149,00
Each
Chemical Identifiers
CAS2082-84-0
IUPAC Namedecyltrimethylazanium bromide
Molecular FormulaC13H30BrN
InChI KeyPLMFYJJFUUUCRZ-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Bromide)≥97.5 to ≤102.5% (Non-U.S. specification)
FormPowder and/or chunks
Assay from Suppliers CofA≥97.5 to ≤102.5% (titration : U.S. specification)
Appearance (Color)White to pale brown
CommentSpecification differs for U.S. and non-U.S. material where indicated
(1-Decyl)trimethylammonium bromide is used in high performance liquid chromatography and in nuclear magnetic resonance. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1-Decyl)trimethylammonium bromide is used in high performance liquid chromatography and in nuclear magnetic resonance. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under inert gas. It is hygroscopic in nature. Incompatible with strong oxidizing agents.
RUO – Research Use Only