(4-Cyanobenzyl)triphenylphosphonium chloride, 99%
(4-Cyanobenzyl)triphenylphosphonium chloride, 99%
(4-Cyanobenzyl)triphenylphosphonium chloride, 99%
Thermo Scientific Chemicals

(4-Cyanobenzyl)triphenylphosphonium chloride, 99%

CAS: 20430-33-5 | C26H21ClNP | 413.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L12187.14
also known as L12187-14
Price (EUR)
121,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
121,00
Each
Chemical Identifiers
CAS20430-33-5
IUPAC Name[(4-cyanophenyl)methyl]triphenylphosphanium chloride
Molecular FormulaC26H21ClNP
InChI KeyGXMFPJIKPZDNCS-UHFFFAOYSA-M
SMILES[Cl-].N#CC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Titration ex Chloride)≥98.5 to ≤101.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only