1-Dodecylamine hydrochloride, 97%
1-Dodecylamine hydrochloride, 97%
1-Dodecylamine hydrochloride, 97%
Thermo Scientific Chemicals

1-Dodecylamine hydrochloride, 97%

CAS: 929-73-7 | C12H28ClN | 221.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L12314.18
also known as L12314-18
Price (TWD)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS929-73-7
IUPAC Namedodecan-1-aminium chloride
Molecular FormulaC12H28ClN
InChI KeyTWFQJFPTTMIETC-UHFFFAOYSA-N
SMILES[Cl-].CCCCCCCCCCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (Argentometric Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only