Thermo Scientific Chemicals

Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin, Thermo Scientific Chemicals

Catalog number: L12708.0B
1000 g, Each
Thermo Scientific Chemicals

Diacetin, mixed isomers, tech. ca 50%, remainder triacetin and monoacetin, Thermo Scientific Chemicals

Catalog number: L12708.0B
1000 g, Each
Quantity
Catalog number: L12708.0B
also known as L12708-0B
Price (USD)
Price: 91.80
Online price: 72.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25395-31-7
IUPAC Name
1-(acetyloxy)-3-hydroxypropan-2-yl acetate
Molecular Formula
C7H12O5
InChI Key
UXDDRFCJKNROTO-UHFFFAOYNA-N
SMILES
CC(=O)OCC(CO)OC(C)=O
Assay (GC)
>45.0% (main component)
Appearance (Color)
Clear, colorless
Form
Liquid
Refractive Index
1.4380-1.4480 @ 20°C

Description

Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diacetin is widely used as plasticizer. It has been used to design and evaluate gliclazide push-pull osmotic pump (PPOP) coated with aqueous colloidal polymer dispersions.

Solubility
Fully miscible in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text