Thermo Scientific Chemicals

Thiophene-2-carboxylic anhydride, 95%, Thermo Scientific Chemicals

Catalog number: L12851.03
1 g, Each
Thermo Scientific Chemicals

Thiophene-2-carboxylic anhydride, 95%, Thermo Scientific Chemicals

Catalog number: L12851.03
1 g, Each
Quantity
Catalog number: L12851.03
also known as L12851-03
Price (USD)
Price: 55.70
Online price: 47.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25569-97-5
IUPAC Name
thiophene-2-carbonyl thiophene-2-carboxylate
Molecular Formula
C10H6O3S2
InChI Key
BKBCDDYQJQGMNX-UHFFFAOYSA-N
SMILES
O=C(OC(=O)C1=CC=CS1)C1=CC=CS1
Assay (GC)
≥94.0%
Appearance (Color)
White to cream to pale yellow
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
52.0-64.0?C

Description

Thiophene-2-carboxylic anhydride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-carboxylic anhydride is used as an organic chemical synthesis intermediate.

Solubility
It hydrolyzes in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text