4-(1-Bromoethyl)benzoic acid, 98%
4-(1-Bromoethyl)benzoic acid, 98%
4-(1-Bromoethyl)benzoic acid, 98%
Thermo Scientific Chemicals

4-(1-Bromoethyl)benzoic acid, 98%

CAS: 113023-73-7 | C9H9BrO2 | 229.073 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 L13025.06
亦稱為 L13025-06
價格 (TWD)
-
Quantity:
5 g
Request bulk or custom format
化學識別
CAS113023-73-7
IUPAC Name4-(1-bromoethyl)benzoic acid
Molecular FormulaC9H9BrO2
InChI KeyVICCYULHZWEWMB-UHFFFAOYNA-N
SMILESCC(Br)C1=CC=C(C=C1)C(O)=O
檢視更多
規格Specification Sheet規格表
Melting Point (clear melt)147.0-156.0°C
FormCrystals or powder or crystalline powder and/or lumpy solid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Appearance (Color)White
4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(1-Bromoethyl)benzoic acid acts as a reagent to prepare 4-(1-bromo-ethyl)-benzoic acid ethyl ester. It is also used as an intermediate for active pharmaceutical ingredient as well as in chemical synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Vandamme, D.; Eyley, S.; den Mooter, G. V.; Muylaert, K.; Thielemans, W. Highly charged cellulose-based nanocrystals as flocculants for harvesting Chlorella vulgaris. Biores. Technol. 2015, 194, 270-275.
  2. Eyley, S.; Thielemans, W. Surface modification of cellulose nanocrystals. Nanoscale 2014, 6, 7764-7779.