Thermo Scientific Chemicals

Dibenzoyl peroxide, 97% (dry wt.), wet with 25% water, Thermo Scientific Chemicals

Catalog number: L13174.0B
1000 g, Each
Thermo Scientific Chemicals

Dibenzoyl peroxide, 97% (dry wt.), wet with 25% water, Thermo Scientific Chemicals

Catalog number: L13174.0B
1000 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L13174.0B
also known as L13174-0B
Price (USD)
Price: 137.00
Your price:
Quantity
-

Chemical Identifiers

CAS
94-36-0
IUPAC Name
benzoyl benzenecarboperoxoate
Molecular Formula
C14H10O4
InChI Key
OMPJBNCRMGITSC-UHFFFAOYSA-N
SMILES
O=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Form
Wet powder or granules
Assay from Supplier's CofA
≥72 to ≤78%
Comment
Contact with metal may produce violent decomposition
Appearance (Color)
White

Description

Benzoyl peroxide is widely utilized as a radical initiator to induce polymerizations. It finds applications for acne treatment, for bleaching flour, hair and teeth and for cross-linking polyester resins. It also has major applications in antiseptic and bleaching properties. It serves as a catalyst for polyester thermoset resins and as a hardener to start the polymerization process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoyl peroxide is widely utilized as a radical initiator to induce polymerizations. It finds applications for acne treatment, for bleaching flour, hair and teeth and for cross-linking polyester resins. It also has major applications in antiseptic and bleaching properties. It serves as a catalyst for polyester thermoset resins and as a hardener to start the polymerization process.

Solubility
Soluble in ether and chloroform. Slightlysoluble in ethanol. Insoluble in water.

Notes
Incompatible with strong oxidizer, reducing agents, acids, bases, alcohols and metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text