L-Lactic acid, anhydrous, 98%
L-Lactic acid, anhydrous, 98%
L-Lactic acid, anhydrous, 98%
Thermo Scientific Chemicals

L-Lactic acid, anhydrous, 98%

CAS: 79-33-4 | C3H6O3 | 90.078 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L13242.22
also known as L13242-22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS79-33-4
IUPAC Name(2S)-2-hydroxypropanoic acid
Molecular FormulaC3H6O3
InChI KeyJVTAAEKCZFNVCJ-REOHCLBHSA-N
SMILESC[C@H](O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)<1.0%
Melting Point49-56?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Useful chiral synthon, building block for depsipeptides.L-Lactic acid is an intermediate in the fermentation of sugar. It plays a vital role in several biochemical processes. It is a major ingredient in processed foods such as yogurt, cottage cheeses, koumiss and kombucha. It gives the sour flavor to sourdough breads. It finds application in pharmaceuticals to prepare water-soluble lactates, topical preparations and cosmetics. It acts as a food preservative, flavoring agent, curing agent, descaler, soap-scum remover and an anti-bacterial agent. It is an active component of lactated Ringer′s solution and Hartmann′s solution. Further, it is used for fluid resuscitation after blood loss due to trauma and surgery.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong bases, strong oxidizing agents, strong reducing agents and metals.
RUO – Research Use Only

General References:

  1. Lv, X.; Guo, Y.; Zhuang, Y.; Wang, Y. Optimization and validation of an extraction method and HPAEC-PAD for determination of residual sugar composition in l-lactic acid industrial fermentation broth with a high salt content. Anal. Methods 2015, 7 (21), 9076-9083.
  2. Yamada, S.; Takasu, A.; Takayama, S.; Kawamura, K. Microwave-assisted solution polycondensation of L-lactic acid using a Dean-Stark apparatus for a non-thermal microwave polymerization effect induced by the electric field. Polym. Chem. 2014, 5 (18), 5283-5288.