Thermo Scientific Chemicals

Sodium cyanide, granules, 98%, Thermo Scientific Chemicals

Catalog number: L13278.0B
1000 g, Each
Thermo Scientific Chemicals

Sodium cyanide, granules, 98%, Thermo Scientific Chemicals

Catalog number: L13278.0B
1000 g, Each
Quantity

Chemical Identifiers

CAS
143-33-9
IUPAC Name
sodium iminomethanide
Molecular Formula
CNNa
InChI Key
MNWBNISUBARLIT-UHFFFAOYSA-N
SMILES
[Na+].[C-]#N
Appearance (Color)
White to pale cream
Form
Granules
Assay from Supplier's CofA
≥97.5%

Description

Sodium cyanide is used as a starting material for the preparation of Reissert compounds and cyanogen bromide, cyanuric chloride and cyanogen chloride. It acts as a catalyst for the aminolysis of esters to primary amides. Furthermore, it is used for fumigation, electroplating, extracting gold and silver in mining industry. In organic synthesis, it is involved in the cyanation reaction of alkyl halides under phase transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cyanide is used as a starting material for the preparation of Reissert compounds and cyanogen bromide, cyanuric chloride and cyanogen chloride. It acts as a catalyst for the aminolysis of esters to primary amides. Furthermore, it is used for fumigation, electroplating, extracting gold and silver in mining industry. In organic synthesis, it is involved in the cyanation reaction of alkyl halides under phase transfer conditions.

Solubility
Soluble in water, ammonia, methanol and ethanol. Slightly soluble in dimethylformamide. Insoluble in dimethylsulphoxide.

Notes
Hygroscopic. Do not store near acids. Incompatible with strong oxidizing agents and carbon dioxide.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text