Xylenes, mixed, 97+%
Xylenes, mixed, 97+%
Xylenes, mixed, 97+%
Thermo Scientific Chemicals

Xylenes, mixed, 97+%

CAS: 1330-20-7 | C8H10
Have Questions?
變更視圖buttonViewtableView
Quantity:
500 mL
1000 mL
2500 mL
產品號碼 L13317.AP
亦稱為 L13317-AP
價格 (TWD)
-
Quantity:
500 mL
Request bulk or custom format
化學識別
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
檢視更多
規格Specification Sheet規格表
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.0% (sum of isomers)
Refractive Index1.4940-1.5000 @ 20?C
Identification (FTIR)Conforms
Xylene is used as a solvent in pharmaceutical, printing, rubber and leather industries. It is an important component of ink, rubber and adhesives as well as thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Xylene is used as a solvent in pharmaceutical, printing, rubber and leather industries. It is an important component of ink, rubber and adhesives as well as thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

Solubility
Miscible with aromatic hydrocarbons. Immiscible with water.

Notes
Contains a mixture of the ortho, meta, and para isomers of xylene. May also contain ethylbenzene. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ferrari-Lima, A. M.; De Souza, R. P.; Mendes, S. S.; Marques, R. G.; Gimenes, M. L.; Machado, N. R. C. F. Photodegradation of benzene, toluene and xylenes under visible light applying N-doped mixed TiO2 and ZnO catalysts. Catal. Today 2015, 241, 40-46.
  2. Nassimbeni, L. R.; Bathori, N. B.; Patel, L. D.; Su, H.; Weber, E. Separation of xylenes by enclathration. Chem. Commun. 2015, 51 (17), 3627-3629.