3,5,5-Trimethylhexanoic acid, 97%
3,5,5-Trimethylhexanoic acid, 97%
3,5,5-Trimethylhexanoic acid, 97%
3,5,5-Trimethylhexanoic acid, 97%
Thermo Scientific Chemicals

3,5,5-Trimethylhexanoic acid, 97%

CAS: 3302-10-1 | C9H18O2 | 158.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Catalog number L13360.AP
also known as L13360-AP
Price (EUR)
137,00
Each
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
137,00
Each
Add to cart
Chemical Identifiers
CAS3302-10-1
IUPAC Name3,5,5-trimethylhexanoic acid
Molecular FormulaC9H18O2
InChI KeyOILUAKBAMVLXGF-UHFFFAOYNA-N
SMILESCC(CC(O)=O)CC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (Silylated GC)≥96.0%
Identification (FTIR)Conforms
Refractive Index1.4265-1.4315 @ 20?C
View more
3,5,5-Trimethylhexanoic acid, is an important organic intermediate. It is also used as an an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5,5-Trimethylhexanoic acid, is an important organic intermediate. It is also used as an an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Miscible with the usual organic solvents.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. JS Preston. The selective solvent extraction of cadmium by mixtures of carboxylic acids and trialkylphosphine sulphides. Part 1. The origin and scope of the synergistic effect. Hydrometallurgy. 1994, 36,(1), 61-78.
  2. LW Reeves.Studies of hydrogen bonding in carboxylic acids. Transactions of the Faraday Society. 1959, 55,(1), 1684-1688.