2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, 98%
2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, 98%
2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, 98%
Thermo Scientific Chemicals

2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate, 98%

CAS: 105832-38-0 | C9H16BF4N3O3 | 301.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L13538.06
also known as L13538-06
Price (USD)
111.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
111.00
Each
Chemical Identifiers
CAS105832-38-0
IUPAC Name[(dimethylamino)[(2,5-dioxopyrrolidin-1-yl)oxy]methylidene]dimethylazanium; tetrafluoroboranuide
Molecular FormulaC9H16BF4N3O3
InChI KeyYEBLHMRPZHNTEK-UHFFFAOYSA-N
SMILESF[B-](F)(F)F.CN(C)C(ON1C(=O)CCC1=O)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or needles
Assay (HPLC)≥97.5%
Appearance (Color)White to pale cream
Melting Point (clear melt)194-201?C
2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate (TSTU) acts as an efficient coupling agent through rapid activation of carboxylic acids to form N-succinimidyl active esters, which react with primary amines to form carboxamides. It is effective even in the presence of water. TSTU is selective towards amines in the presence of alcoholic group. It is employed as a coupling regent in the synthesis of peptides. With amine-protected alpha-amino acids, it brings about coupling without significant epimerization. It is useful for linking of sensor molecules with amino groups to amino-functionalized AFM (atomic force microscope) tips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Succinimido-1,1,3,3-tetramethyluronium tetrafluoroborate (TSTU) acts as an efficient coupling agent through rapid activation of carboxylic acids to form N-succinimidyl active esters, which react with primary amines to form carboxamides. It is effective even in the presence of water. TSTU is selective towards amines in the presence of alcoholic group. It is employed as a coupling regent in the synthesis of peptides. With amine-protected alpha-amino acids, it brings about coupling without significant epimerization. It is useful for linking of sensor molecules with amino groups to amino-functionalized AFM (atomic force microscope) tips.

Solubility
Soluble in acetonitrile, DMF, chloroform, dichlorormethane and acetonitrile.

Notes
Moisture sensitive, Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Yang, Q.; Wang, H.; Lan, X.; Cheng, B.; Chen, S.; Shi, H.; Xiao, H.; Ma, Y. Reflection-mode micro-spherical fiber-optic probes for in vitro real-time and single-cell level pH sensing. Sens. Actuators B 2015, 207, 571-580.
  2. Yang, Q.; Wang, H.; Chen, S.; Lan, X.; Xiao, H.; Shi, H.; Ma, Y. Fiber-Optic-Based Micro-Probe Using Hexagonal 1-in-6 Fiber Configuration for Intracellular Single-Cell pH Measurement. Anal. Chem. 2015, 87 (14), 7171-7179.