Tris(3,6-dioxaheptil)amina, 95 %, Thermo Scientific Chemicals
Tris(3,6-dioxaheptil)amina, 95 %, Thermo Scientific Chemicals
Tris(3,6-dioxaheptil)amina, 95 %, Thermo Scientific Chemicals
Tris(3,6-dioxaheptil)amina, 95 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tris(3,6-dioxaheptil)amina, 95 %, Thermo Scientific Chemicals

CAS: 70384-51-9 | C15H33NO6 | 323.43 g/mol
Have Questions?
Número de catálogoCantidad
L13544.065 g
Número de catálogo L13544.06
Precio (USD)
-
Cantidad:
5 g
Pedido a granel o personalizado
Identificadores químicos
CAS70384-51-9
IUPAC Name8-[2-(2-methoxyethoxy)ethyl]-2,5,11,14-tetraoxa-8-azapentadecane
Molecular FormulaC15H33NO6
InChI KeyXGLVDUUYFKXKPL-UHFFFAOYSA-N
SMILESCOCCOCCN(CCOCCOC)CCOCCOC
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
FormLiquid
Refractive Index1.4465-1.4490 @ 20?C
Appearance (Color)Clear pale yellow to yellow to orange to brown
Assay (GC)≥94.0%
Tris(3,6-dioxaheptyl)amine acts as a phase transfer catalyst used in the preparation of 1,3-dideaza-2'-deoxyadenosine and related benzimidazole 2'-deoxyribonucleosides. It is also used as an intermediate in the synthesis of telithromycin. Further, it is used in the oxidation of arylmethanols under phase-transfer conditions. In addition to this, it acts as a cryptand during phase-transfer glycosylation of 2-amino-4-methoxy-7H-pyrrolo[2,3-d]pyrimidine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Aplicaciones
la tris(3,6-dioxaheptil)amina actúa como catalizador de transferencia de fase utilizado en la preparación de 1,3-dideaza-2′-desoxiadenosina y benzimidazol 2′-desoxirribonucleósidos relacionados. También se utiliza como intermedio en la síntesis de telitromicina. Además, se emplea en la oxidación de los arilmetanoles en condiciones de transferencia de fase. Además de esto, actúa como criptando durante la glicosilación de transferencia de fase 2-amino-4-metoxi-7H-pirrollo[2,3-d]pirimidina.

Solubilidad
Miscible con agua.

Notas
Sensible al aire e higroscópico. Incompatible con agentes oxidantes fuertes y ácidos fuertes.
RUO – Research Use Only

General References:

  1. Acyclic solid-liquid phase-transfer catalyst (see Appendix 2).
  2. Solubilizes salts of alkali metals and of transition metals, e.g. PdCl2, RuCl3 and RhCl3, allowing these to be used in hydrogenation reactions: Synth. Commun., 19, 2833 (1989).
  3. Forms isolable 2:1 complexes with Grignard reagents: Tetrahedron, 45, 171 (1989).
  4. Accelerates dehydrobromination of alkyl halides in the presence of KOH: Org. Synth. Coll., 9, 539 (1998). For use in the Wittig reaction, see Cyclopropyl triphenyl phosphonium bromide, B25102.
  5. Has been used in a number of aromatic nucleophilic substitution reactions:
  6. In studies of solvent-free SNAr reactions, TDA-1 was found to be the best catalyst: Synth. Commun., 20, 2855 (1990); Heterocycles, 32, 1947 (1991). In combination with CuCl, is also a useful catalyst in the Ullmann aryl ether synthesis: J. Org. Chem., 50, 3717 (1985). An improved N-arylation of amides utilizes a catalyst combination of CuCl and TDA-1 (5:1): Synthesis, 312 (1989).
  7. For use in a stereospecific glycosylation reaction of a pyrimidine base, see: Helv. Chim. Acta, 71, 1573 (1988).
  8. Quick, M. P.; Wünsch, B. Synthesis of hydrogenated 2-benzazepin-1-ones by the addition of aryllithium intermediates to isocyanates. Tetrahedron: Asymmetry 2015, 26 (5-6), 276-280.
  9. Li, H.; Pang, Z.; Gao, P.; Wang, L. Fe(III)-catalyzed grafting copolymerization of lignin with styrene and methyl methacrylate through AGET ATRP using triphenyl phosphine as a ligand. RSC Adv. 2015, 5 (67), 54387-54394.