3-Chlorothiophene-2-carboxylic acid, 97+%
3-Chlorothiophene-2-carboxylic acid, 97+%
3-Chlorothiophene-2-carboxylic acid, 97+%
Thermo Scientific Chemicals

3-Chlorothiophene-2-carboxylic acid, 97+%

CAS: 59337-89-2 | C5H3ClO2S | 162.587 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L13560.03
also known as L13560-03
Price (EUR)
44,10
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
44,10
Each
Chemical Identifiers
CAS59337-89-2
IUPAC Name3-chlorothiophene-2-carboxylate
Molecular FormulaC5H2ClO2S
InChI KeyBXEAAHIHFFIMIE-UHFFFAOYSA-M
SMILES[O-]C(=O)C1=C(Cl)C=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow to pale brown
FormPowder or crystalline powder
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
Assay (Silylated GC)≥97.0%
Identification (FTIR)Conforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only