Thermo Scientific Chemicals

Filter aid, Harborlite 800, Thermo Scientific Chemicals

Catalog number: L13608.36
500 g, Each
Thermo Scientific Chemicals

Filter aid, Harborlite 800, Thermo Scientific Chemicals

Catalog number: L13608.36
500 g, Each
Quantity
Catalog number: L13608.36
also known as L13608-36
Price (JPY)

Chemical Identifiers

CAS
130885-09-5
Comment
Permeability: 1.40-2.40 Darcy
Appearance (Color)
White to pale grey
Comment
Cake density or Wet density (by centrifugation): 175 - 205 g/litre
Comment
Flotation max: < 15% by volume
Form
Powder

Description

Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter aid, Harborlite 800 is used in multiple industrial applications such as chemical processing, mineral and lubrication oil production, biodiesel purification. It is also used for filtration performance in brewing, juices, wine, sugar syrups, edible vegetable and fish oils.

Solubility
Insoluble in water.

Notes
Harborlite is a registered trademark of Harborlite Corportion.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text