Thermo Scientific Chemicals

3-Methyl-1-butanol, mixture of isomers, 99%, Thermo Scientific Chemicals

Catalog number: L13660.AE
100 mL, Each
Thermo Scientific Chemicals

3-Methyl-1-butanol, mixture of isomers, 99%, Thermo Scientific Chemicals

Catalog number: L13660.AE
100 mL, Each
Quantity
Catalog number: L13660.AE
also known as L13660-AE
Price (USD)
Price: 36.10
Online price: 31.65
Your price:
Quantity
-

Chemical Identifiers

CAS
123-51-3
IUPAC Name
3-methylbutan-1-ol
Molecular Formula
C5H12O
InChI Key
PHTQWCKDNZKARW-UHFFFAOYSA-N
SMILES
CC(C)CCO
Assay (GC)
≥98.5%
Form
Liquid
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Refractive Index
1.4055-1.4090 @ 20?C

Description

3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac′s reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.

Solubility
Miscible with water, alcohol, ether, benzene, chloroform, petroleum ether, glacial acetic acid, acetone and oils.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides and acid anhydrides.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text