Magnesium sulfate, dried, contains ca 1-2 mol water of hydration
Magnesium sulfate, dried, contains ca 1-2 mol water of hydration
Magnesium sulfate, dried, contains ca 1-2 mol water of hydration
Thermo Scientific Chemicals

Magnesium sulfate, dried, contains ca 1-2 mol water of hydration

CAS: 22189-08-8 | MgO4S | 120.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number L13739.22
also known as L13739-22
Price (EUR)
22,70
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
22,70
Each
Chemical Identifiers
CAS22189-08-8
IUPAC Namemagnesium(2+) sulfate
Molecular FormulaMgO4S
InChI KeyCSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (unspecified)76-78%
CommentWaters of Hydration : ≤2.3 mol
FormPowder
Suitable for the preliminary drying of organic liquids, for example solvent extracts from reaction workups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only