Urea hydrogen peroxide adduct, 97%
Urea hydrogen peroxide adduct, 97%
Urea hydrogen peroxide adduct, 97%
Urea hydrogen peroxide adduct, 97%
Thermo Scientific Chemicals

Urea hydrogen peroxide adduct, 97%

CAS: 124-43-6 | CH6N2O3 | 94.07 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 L13940.22
亦稱為 L13940-22
價格 (TWD)
-
申請報價
Quantity:
100 g
Request bulk or custom format
化學識別
CAS124-43-6
IUPAC Nameperoxol; urea
Molecular FormulaCH6N2O3
InChI KeyAQLJVWUFPCUVLO-UHFFFAOYSA-N
SMILESOO.NC(N)=O
檢視更多
規格Specification Sheet規格表
Melting Point (clear melt)87-93?C
FormCrystals or crystalline powder or tables
Identification (FTIR)Conforms
CommentAvailable oxygen 16-18%
Appearance (Color)White
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Urea hydrogen peroxide is an antiseptic, disinfectant and bleaching agent used in pharmaceuticals and cosmetics respectively. It is used in the whitening of teeth and relieves minor inflammation of gums, oral mucosal surfaces and lips. It finds application in the preparation of plastics, blue print development and starch modification. In addition to this, it is used as a source of hydrogen peroxide easily handled in the laboratory.

Solubility
Soluble in water.

Notes
Incompatible with acetic anhydride, aniline and carboxylic acids.
RUO – Research Use Only

General References:

  1. Useful, low-hazard alternative to high-concentration H2O2 in epoxidations, Baeyer-Villiger reactions, N-oxidations, etc.: Synlett, 533 (1990). Alkenes can be epoxidized in high yield in 1,1,1,3,3,3-Hexafluoro-2-propanol, A12747 as solvent: Eur. J. Org. Chem., 3290 (2002). For use in combination with N,N'-Dicyclohexyl carbodiimide, A10973 , in epoxidation of alkenes, see: J. Org. Chem., 63, 1730 (1998); in combination with Maleic anhydride, A12178 , for epoxidation and Baeyer-Villiger reactions: Heterocycles, 36, 1075 (1993). For an extensive review of the Baeyer-Villiger reaction, see: Org. React., 43, 251 (1993).
  2. For use in partial oxidation of nitriles to amides, see: Synth. Commun., 23, 3149 (1993).
  3. In combination with phthalic anhydride is an effective mild and safe oxidant for N-heteroaromatic compounds and tertiary amines to N-oxides: Chem. Ber., 125, 1965 (1992)
  4. Can be used to generate the powerful oxidant peroxytrifluoroacetic acid from TFAA, avoiding the use of hazardous 90% H2O2: Tetrahedron Lett., 33, 4835 (1992). The same system has also been reported for oxidation of electron-deficient pyridines to the N-oxides: Tetrahedron Lett., 41, 2299 (2000). Similarly, performic acid can be generated in situ, providing a mild system for oxidation of aromatic aldehydes to benzoic acids: Synth. Commun., 31, 2195 (2001). See also Sodium percarbonate, A16045 .
  5. Yazu, K.; Takanohashi, T. Oxidative Desulfurization of Coal Tar Pitch Using a Urea-Hydrogen Peroxide Complex/Carboxylic Anhydride System in THF. Chem. Lett. 2015, 44 (2), 169-170.
  6. Nikoorazm, M.; Ghorbani-Choghamarani, A.; Ghorbani, F.; Mahdavi, H.; Karamshahi, Z. Bidentate salen Cu(II) complex functionalized on mesoporous MCM-41 as novel nano catalyst for the oxidative coupling of thiols into disulfides using urea hydrogen peroxide (UHP). J. Porous Mater. 2015, 22 (1), 261-267.